Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydrazino-3-(pyrrolidin-1-ylsulfonyl)pyridine ≥95%

Supplier: MATRIX SCIENTIFIC MS

115171-500MG 115171-1G
102895-512EA 693.85 USD InStock
102895-512 102895-514
2-Hydrazino-3-(pyrrolidin-1-ylsulfonyl)pyridine ≥95%
2-Hydrazino-3-(pyrrolidin-1-ylsulfonyl)pyridine
Hazard: Irritant


Formula: C₉H₁₄N₄O₂S
MDL Number: MFCD12174504
CAS Number: 1155916-50-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty