Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Acetylamino)-3-(3,5-difluorophenyl)-2-propenoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

115260-500MG 115260-1G
102893-392EA 412.31 USD InStock
102893-392 102893-394
2-(Acetylamino)-3-(3,5-difluorophenyl)-2-propenoic acid ≥95%
2-(Acetylamino)-3-(3,5-difluorophenyl)-2-propenoic acid
Hazard: Irritant


Formula: C₁₁H₉F₂NO₃
MDL Number: MFCD27680350
CAS Number: 883035-24-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty