Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-(trifluoromethyl)pyrazine ≥97%

Supplier: MATRIX SCIENTIFIC MS

115275-1G 115275-5G
102894-524EA 250.77 USD InStock
102894-524 102894-622
2-Chloro-6-(trifluoromethyl)pyrazine ≥97%
2-Chloro-6-(trifluoromethyl)pyrazine
Hazard: Corrosive/Refrigerate


Formula: C₅H₂ClF₃N₂
MDL Number: MFCD13189477
CAS Number: 61655-69-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty