Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Trifluoromethyl)cyclopropanemethanol ≥95%

Supplier: MATRIX SCIENTIFIC MS

115428-100MG 115428-250MG 115428-500MG
102896-156EA 1318.46 USD InStock
102896-156 102896-158 102896-160
1-(Trifluoromethyl)cyclopropanemethanol ≥95%
1-(Trifluoromethyl)cyclopropanemethanol
Hazard: Irritant


Formula: C₅H₇F₃O
MDL Number: MFCD16990760
CAS Number: 371917-17-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty