Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-3-(trifluoromethyl)piperidin-4-one ≥95%

Supplier: MATRIX SCIENTIFIC MS

115458-250MG 115458-500MG 115458-1G
102896-310EA 3366.15 USD InStock
102896-310 102896-312 102896-332
1-Benzyl-3-(trifluoromethyl)piperidin-4-one ≥95%
1-Benzyl-3-(trifluoromethyl)piperidin-4-one
Hazard: Irritant


Formula: C₁₃H₁₄F₃NO
MDL Number: MFCD21601941
CAS Number: 373604-45-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty