Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-3-(piperidin-4-yl)-1H-indazole hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

115487-250MG 115487-500MG 115487-1G
102896-484EA 1855.38 USD InStock
102896-484 102896-486 102896-488
6-Fluoro-3-(piperidin-4-yl)-1H-indazole hydrochloride ≥95%
6-Fluoro-3-(piperidin-4-yl)-1H-indazole hydrochloride
Hazard: Irritant


Formula: C₁₂H₁₅ClFN₃
MDL Number: MFCD12912622
CAS Number: 153025-27-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty