Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Azabicyclo[2.1.1]hexane-2,5-dicarboxylic acid-2-tert-butyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

115511-100MG 115511-250MG 115511-1G
102896-564EA 1593.85 USD InStock
102896-564 102896-566 102896-568
2-Azabicyclo[2.1.1]hexane-2,5-dicarboxylic acid-2-tert-butyl ester ≥95%
2-Azabicyclo[2.1.1]hexane-2,5-dicarboxylic acid-2-tert-butyl ester
Hazard: Irritant


Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
CAS Number: 615575-74-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty