Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl-3aR,6aR-octahydropyrrolo[3,2-b]pyrrole-1-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

115517-250MG 115517-500MG 115517-1G
102896-598EA 1647.69 USD InStock
102896-598 102896-600 102896-602
Benzyl-3aR,6aR-octahydropyrrolo[3,2-b]pyrrole-1-carboxylate ≥95%
Benzyl-3aR,6aR-octahydropyrrolo[3,2-b]pyrrole-1-carboxylate
Hazard: Irritant


Formula: C₁₄H₁₈N₂O₂
MDL Number: MFCD18632603
CAS Number: 1251009-14-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty