Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Octahydroisoindole ≥95%

Supplier: MATRIX SCIENTIFIC MS

115552-1G 115552-5G 115552-500MG
102896-808EA 1033.85 USD InStock
102896-808 102896-810 102899-446
cis-Octahydroisoindole ≥95%
cis-Octahydroisoindole
Hazard: Irritant


Formula: C₈H₁₅N
MDL Number: MFCD07778620
CAS Number: 1470-99-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty