Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-1-Boc-3-(methylamino)piperidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

115607-500MG 115607-1G 115607-250MG
102896-962EA 1241.54 USD InStock
102896-962 102896-964 102897-008
(S)-(+)-1-Boc-3-(methylamino)piperidine ≥95%
(S)-(+)-1-Boc-3-(methylamino)piperidine
Hazard: Irritant


Formula: C₁₁H₂₂N₂O₂
MW: 214.309 g/mol
MDL Number: MFCD09752964
CAS Number: 912368-73-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty