Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloropropyl)diphenylsulfonium tetrafluoroborate ≥97%

Supplier: MATRIX SCIENTIFIC MS

115669-1G 115669-5G 115669-25G
102897-154EA 330.77 USD InStock
102897-154 102897-156 102897-158
(3-Chloropropyl)diphenylsulfonium tetrafluoroborate ≥97%
(3-Chloropropyl)diphenylsulfonium tetrafluoroborate
Hazard: Irritant


Formula: C₁₅H₁₆BClF₄S
MW: 350.61 g/mol
MDL Number: MFCD00074932
CAS Number: 33462-80-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty