Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Boc-octahydro-pyrrolo[3,4-c]pyridine ≥95%

Supplier: MATRIX SCIENTIFIC MS

115758-250MG 115758-500MG 115758-1G
102897-420EA 1512.31 USD InStock
102897-420 102897-422 102897-424
5-Boc-octahydro-pyrrolo[3,4-c]pyridine ≥95%
5-Boc-octahydro-pyrrolo[3,4-c]pyridine
Hazard: Irritant


Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD06858491
CAS Number: 1257389-94-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty