Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazolo[3,4-c]pyridine-5-carboxylic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

115801-250MG 115801-500MG 115801-1G
102898-144EA 1512.31 USD InStock
102898-144 102898-146 102898-148
1H-Pyrazolo[3,4-c]pyridine-5-carboxylic acid ≥95%
1H-Pyrazolo[3,4-c]pyridine-5-carboxylic acid
Hazard: Irritant


Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
MDL Number: MFCD18250838
CAS Number: 1256824-45-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty