Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid methyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

115802-250MG 115802-500MG 115802-1G
102898-150EA 1318.46 USD InStock
102898-150 102898-152 102898-154
1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid methyl ester ≥95%
1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid methyl ester
Hazard: Irritant


Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
MDL Number: MFCD18425635
CAS Number: 1352394-18-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty