Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimesic acid trimethyl ester ≥97%

Supplier: MATRIX SCIENTIFIC MS

115916-100G 115916-5G
102898-992EA 678.46 USD InStock
102898-992 102899-876
Trimesic acid trimethyl ester ≥97%
Trimesic acid trimethyl ester
Hazard: Irritant


Formula: C₁₂H₁₂O₆
MW: 252.22
MDL Number: MFCD00008434
CAS Number: 2672-58-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty