Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyldimethyl(2-propynyloxy)silane ≥97%

Supplier: MATRIX SCIENTIFIC MS
Warning

115984-1G 115984-25G
102900-140EA 43.08 USD InStock
102900-140 102900-144
tert-Butyldimethyl(2-propynyloxy)silane ≥97%
tert-Butyldimethyl(2-propynyloxy)silane
Purity: 97% CAS: 76782-82-6 MDL Number: MFCD01863574 Molecular Formula: C9H18OSi Molecular Weight: 170,33 Hazard: Irritant


Formula: C₉H₁₈OSi
MW: 170.33 g/mol
MDL Number: MFCD28156589
CAS Number: 76782-82-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty