Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-amino-5-bromobenzoate ≥97%

Supplier: MATRIX SCIENTIFIC MS

113943-1G 113943-10G 113943-50G
102898-288EA 510.77 USD InStock
102898-288 102898-290 102898-292
Methyl-2-amino-5-bromobenzoate ≥97%
Methyl-2-amino-5-bromobenzoate
Hazard: Irritant


Formula: C₈H₈BrNO₂
MDL Number: MFCD00144761
CAS Number: 52727-57-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty