Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorocyclohexanone ≥97%

Supplier: MATRIX SCIENTIFIC MS

114079-1G 114079-10G
102893-266EA 60 USD InStock
102893-266 102893-268
2-Chlorocyclohexanone ≥97%
2-Chlorocyclohexanone
Hazard: Irritant


Formula: C₆H₉ClO
MW: 132.59 g/mol
Boiling Pt: 203
MDL Number: MFCD00001626
CAS Number: 822-87-7
UN: 3335
ADR: 9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty