Human beta-Amyloid (1-42)
Supplier: Hytest Ltd
AS-21791 AS-21793
102996-168EA 507.15 USD InStock
102996-168 102996-170
Human beta-Amyloid (1-42)
Proteins and Peptides
This is a salt form of Aβ(1-42) known to form aggregates within a few days in water or recommended solvents. The salt form has been found to form fibrils, exerts cytotoxicity in neuronal models and so, used in studies that evaluate activity of agents that reverse amyloid-induced cytotoxicity.
  • Peptide Content: ≥60%
Citations:
Iwasaki, K. et al. (2007). Nilvadipine prevents the impairment of spatial memory induced by cerebral ischemia combined with β-amyloid in rats. Biol Pharm Bull 30, 698.
Iwasaki, K. et al. (2006). Cerebral ischemia combined with β-amyloid impairs spatial memory in the eight-arm radial maze task in rats. Brain Res 1097, 216. doi: 10.1016/j.brainres.2006.04.073.
Hirata, K. et al. (2005). A novel neurotrophic agent, T-817MA [1-{3-[2-(1-Benzothiophen-5-yl) Ethoxy] Propyl}-3-azetidinol Maleate], attenuates amyloid-β-induced neurotoxicity and promotes neurite outgrowth in rat cultured central nervous system neurons. JPET 314, 252. doi: 10.1124/jpet.105.083543.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty