Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Human Beta-Amyloid (1-42), Biotin

Supplier: Hytest Ltd

AS-61484-01 AS-61484-05
103006-508EA 349.31 USD InStock
103006-508 103006-798
Human Beta-Amyloid (1-42), Biotin
Proteins and Peptides
Biotinylated forms of Aβ are used commonly for interaction studies. Studies have revealed that biotinylation of a lysine at the C-terminus or N-terminal biotinylation of the beta-amyloid peptides influences the secondary structure conformation.
This beta-amyloid 1-42 peptide is biotinylated to a lysine residue attached to the C-terminal end.
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA-K(Biotin)-NH2
MW: 4867.6 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty