Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrazolo[1,5-a]pyridin-7-amine

Supplier: MATRIX SCIENTIFIC MS

077100-500MG 077100-1G
102664-464EA 338.46 USD InStock
102664-464 102664-466
Tetrazolo[1,5-a]pyridin-7-amine
Tetrazolo[1,5-a]pyridin-7-amine
Hazard: Irritant


Formula: C₅H₅N₅
MDL Number: MFCD16618482
CAS Number: 1228551-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty