Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-{[2-Chloro-5-(trifluoromethyl)phenyl]amino}-5-oxopentanoic acid

Supplier: MATRIX SCIENTIFIC MS

077149-500MG 077149-1G 077149-5G
102664-726EA 109.23 USD InStock
102664-726 102664-728 102664-730
5-{[2-Chloro-5-(trifluoromethyl)phenyl]amino}-5-oxopentanoic acid
5-{[2-Chloro-5-(trifluoromethyl)phenyl]amino}-5-oxopentanoic acid
Hazard: Irritant


Formula: C₁₂H₁₁ClF₃NO₃
MDL Number: MFCD00029870
CAS Number: 199803-46-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty