Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(1H-pyrazol-1-yl)benzoate

Supplier: MATRIX SCIENTIFIC MS

077429-500MG 077429-1G 077429-5G
102666-198EA 258.46 USD InStock
102666-198 102666-200 102666-202
Methyl-3-(1H-pyrazol-1-yl)benzoate
Methyl-3-(1H-pyrazol-1-yl)benzoate
Hazard: Irritant


Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD20441358
CAS Number: 168618-35-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty