Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminohexanoic acid hydrochloride

Supplier: MATRIX SCIENTIFIC MS

077457-500MG 077457-1G 077457-5G
102666-352EA 646.15 USD InStock
102666-352 102666-354 102666-356
4-Aminohexanoic acid hydrochloride
4-Aminohexanoic acid hydrochloride


Formula: C₆H₁₄ClNO₂
MDL Number: MFCD20441443
CAS Number: 62000-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty