Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-6-oxo-1,6-dihydropyrimidine-5-carbonitrile

Supplier: MATRIX SCIENTIFIC MS

077587-500MG 077587-1G
102667-010EA 516.92 USD InStock
102667-010 102667-012
1-Methyl-6-oxo-1,6-dihydropyrimidine-5-carbonitrile
1-Methyl-6-oxo-1,6-dihydropyrimidine-5-carbonitrile
Hazard: Irritant


Formula: C₆H₅N₃O
MDL Number: MFCD19982746
CAS Number: 1330756-29-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty