Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(3-methyl-4-nitro-1H-pyrazol-1-yl)propanoate

Supplier: MATRIX SCIENTIFIC MS

080344-500MG 080344-1G
102669-206EA 516.92 USD InStock
102669-206 102669-208
Methyl-3-(3-methyl-4-nitro-1H-pyrazol-1-yl)propanoate
Methyl-3-(3-methyl-4-nitro-1H-pyrazol-1-yl)propanoate
Hazard: Irritant


Formula: C₈H₁₁N₃O₄
MDL Number: MFCD04969163
CAS Number: 1001499-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty