Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-[(5-amino-1,3,4-thiadiazol-2-yl)thio]-3-oxobutanoate

Supplier: MATRIX SCIENTIFIC MS

080535-500MG 080535-1G 080535-5G
102670-026EA 258.46 USD InStock
102670-026 102670-028 102670-030
Ethyl-4-[(5-amino-1,3,4-thiadiazol-2-yl)thio]-3-oxobutanoate
Ethyl-4-[(5-amino-1,3,4-thiadiazol-2-yl)thio]-3-oxobutanoate
Hazard: Irritant


Formula: C₈H₁₁N₃O₃S₂
MDL Number: MFCD00109373
CAS Number: 220088-59-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty