Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyl-3-oxo-2-[(3-oxo-5-phenylcyclohex-1-en-1-yl)oxy]butanoate

Supplier: MATRIX SCIENTIFIC MS

080625-500MG 080625-1G
102670-452EA 193.85 USD InStock
102670-452 102670-454
Isopropyl-3-oxo-2-[(3-oxo-5-phenylcyclohex-1-en-1-yl)oxy]butanoate
Isopropyl-3-oxo-2-[(3-oxo-5-phenylcyclohex-1-en-1-yl)oxy]butanoate
Hazard: Irritant


Formula: C₁₉H₂₂O₅
MDL Number: MFCD22374929
CAS Number: 1374509-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty