Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,7-Dimethoxy-3,4-dihydroisoquinoline-2(1H)-carboximidamide sulfate

Supplier: MATRIX SCIENTIFIC MS

080810-500MG 080810-1G 080810-5G
102671-336EA 258.46 USD InStock
102671-336 102671-338 102671-340
6,7-Dimethoxy-3,4-dihydroisoquinoline-2(1H)-carboximidamide sulfate
6,7-Dimethoxy-3,4-dihydroisoquinoline-2(1H)-carboximidamide sulfate
Hazard: Irritant


Formula: C₁₂H₁₉N₃O₆S
Melting Pt: 261…262 °C
MDL Number: MFCD11983184
CAS Number: 1179369-64-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty