Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Diphenylmethyl)piperazine-1-carboximidamide sulfate

Supplier: MATRIX SCIENTIFIC MS

080824-500MG 080824-1G 080824-5G
102671-416EA 193.85 USD InStock
102671-416 102671-418 102671-420
4-(Diphenylmethyl)piperazine-1-carboximidamide sulfate
4-(Diphenylmethyl)piperazine-1-carboximidamide sulfate
Hazard: Irritant


Formula: C₁₈H₂₄N₄O₄S
MW: 392.48 g/mol
MDL Number: MFCD22421994
CAS Number: 1194681-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty