Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorophenyl)piperazine-1-carboximidamide sulfate

Supplier: MATRIX SCIENTIFIC MS

080828-500MG 080828-1G 080828-5G
102671-440EA 193.85 USD InStock
102671-440 102671-442 102671-444
4-(4-Chlorophenyl)piperazine-1-carboximidamide sulfate
4-(4-Chlorophenyl)piperazine-1-carboximidamide sulfate
Hazard: Irritant


Formula: C₁₁H₁₇ClN₄O₄S
MDL Number: MFCD01748419
CAS Number: 1417568-42-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty