Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Methoxyphenyl)piperazine-1-carboximidamide sulfate

Supplier: MATRIX SCIENTIFIC MS

080829-500MG 080829-1G 080829-5G
102671-446EA 193.85 USD InStock
102671-446 102671-448 102671-450
4-(4-Methoxyphenyl)piperazine-1-carboximidamide sulfate
4-(4-Methoxyphenyl)piperazine-1-carboximidamide sulfate
Hazard: Irritant


Formula: C₁₂H₂₀N₄O₅S
MDL Number: MFCD22421998
CAS Number: 1417567-67-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty