Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl [2-(cyclopropylamino)-6-oxo-1,4,5,6-tetrahydropyrimidin-5-yl]acetate

Supplier: MATRIX SCIENTIFIC MS

080869-500MG 080869-1G 080869-5G
102671-660EA 387.69 USD InStock
102671-660 102671-662 102671-664
Methyl [2-(cyclopropylamino)-6-oxo-1,4,5,6-tetrahydropyrimidin-5-yl]acetate
Methyl [2-(cyclopropylamino)-6-oxo-1,4,5,6-tetrahydropyrimidin-5-yl]acetate
Hazard: Irritant


Formula: C₁₀H₁₅N₃O₃
MDL Number: MFCD22374983
CAS Number: 1379811-77-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty