Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-methylhydrazinecarboxylate

Supplier: MATRIX SCIENTIFIC MS

080939-500MG 080939-1G 080939-5G
102671-992EA 516.92 USD InStock
102671-992 102671-994 102671-996
Ethyl-1-methylhydrazinecarboxylate
Ethyl-1-methylhydrazinecarboxylate
Hazard: Irritant


Formula: C₄H₁₀N₂O₂
MDL Number: MFCD18431358
CAS Number: 760-81-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty