Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichloropyridine methanesulfonate ≥97%

Supplier: MATRIX SCIENTIFIC MS

083835-1G 083835-10G 083835-50G
102840-862EA 170.77 USD InStock
102840-862 102840-864 102840-866
3,4-Dichloropyridine methanesulfonate ≥97%
3,4-Dichloropyridine methanesulfonate
Hazard: Irritant


Formula: C₆H₇Cl₂NO₃S
MDL Number: MFCD08274935
CAS Number: 1215561-47-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty