Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-5-chloro-1H-imidazole ≥97%

Supplier: MATRIX SCIENTIFIC MS

083935-1G 083935-10G
102841-116EA 226.15 USD InStock
102841-116 102841-118
2-Ethyl-5-chloro-1H-imidazole ≥97%
2-Ethyl-5-chloro-1H-imidazole
Hazard: Irritant


Formula: C₅H₇ClN₂
MDL Number: MFCD08437609
CAS Number: 944898-66-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty