Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]1H-1,2,3-triazole-4,5-dicarboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

115133-500MG 115133-1G
102891-762EA 693.85 USD InStock
102891-762 102891-764
Dimethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]1H-1,2,3-triazole-4,5-dicarboxylate ≥95%
Dimethyl-1-[2-(4-fluorophenyl)-2-oxoethyl]1H-1,2,3-triazole-4,5-dicarboxylate


Formula: C₁₄H₁₂FN₃O₅
MDL Number: MFCD28023528
CAS Number: 1610377-03-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty