BBL™ Penicillinase, BD Biosciences
Supplier: Seca Med.Waagen u.Meßsysteme
211898 211897 211899
90000-424PK 612.17 USD InStock
90000-424 90000-422 90000-426
BBL™ Penicillinase, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
This enzyme is specifically used for the inactivation of penicillin, for increasing the susceptibility of Gram-negative bacteria to antibiotics.
  • Interferes with the antigen-antibody reaction
  • Increases susceptibility of Gram-negative bacteria to antibiotics
Penicillinases provide multi-resistance to antibiotics such as penicillins, cephamycins, and carbapenems. Through hydrolysis, penicillinases break the molecular structure that all these antibiotics have in common.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty