Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-DLK1 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 211309]

Supplier: Novus Biologicals
FAB1144G-025 FAB1144G-100
103250-222EA 258.86 USD InStock
103250-222 103250-224
Anti-DLK1 Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 211309]
Antibodies
Type: Primary
Antigen: DLK1
Clonality: Monoclonal
Clone: 211309
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty