Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL23R Mouse Monoclonal Antibody (PerCP (Peridinin-chlorophyll Protein Complex)) [clone: 218213]

Supplier: Novus Biologicals
FAB14001C-025 FAB14001C-100
103250-772EA 302.25 USD InStock
103250-772 103250-774
Anti-IL23R Mouse Monoclonal Antibody (PerCP (Peridinin-chlorophyll Protein Complex)) [clone: 218213]
Antibodies
Type: Primary
Antigen: IL23R
Clonality: Monoclonal
Clone: 218213
Conjugation: PerCP (Peridinin-chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty