Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD8A Rat Monoclonal Antibody (Allophycocyanin) [clone: 53-6.7]

Supplier: Novus Biologicals
FAB116A-025 FAB116A-100
103251-342EA 172.09 USD InStock
103251-342 103251-344
Anti-CD8A Rat Monoclonal Antibody (Allophycocyanin) [clone: 53-6.7]
Antibodies
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Allophycocyanin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty