Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL17RA Mouse Monoclonal Antibody (Allophycocyanin) [clone: 133617]

Supplier: Novus Biologicals
FAB177A-025 FAB177A-100
103251-714EA 331.17 USD InStock
103251-714 103251-716
Anti-IL17RA Mouse Monoclonal Antibody (Allophycocyanin) [clone: 133617]
Antibodies
Type: Primary
Antigen: IL17RA
Clonality: Monoclonal
Clone: 133617
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty