Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CLEC7A Mouse Monoclonal Antibody (Phycoerythrin) [clone: 259931]

Supplier: Novus Biologicals
FAB1859P-025 FAB1859P-100
103251-938EA 258.86 USD InStock
103251-938 103251-940
Anti-CLEC7A Mouse Monoclonal Antibody (Phycoerythrin) [clone: 259931]
Antibodies
Type: Primary
Antigen: CLEC7A
Clonality: Monoclonal
Clone: 259931
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty