BBL™ Prepared Plated Media 7H11 Agar Base, BD
Supplier: Seca Med.Waagen u.Meßsysteme
212203 221392
90000-526EA 342.84 USD InStock
90000-526 90001-352
BBL™ Prepared Plated Media 7H11 Agar Base, BD
Media for Microbiology Dehydrated Media for Microbiology
7H11 Agar is used in qualitative procedures for isolation and cultivation of mycobacteria, especially Mycobacterium tuberculosis, from clinical and nonclinical specimens.
  • Highly selective medium
  • Malachite green inhibits the growth of contaminating bacteria
The addition of antimicrobial agents to 7H11 Agar improves the recovery of mycobacteria from specimens containing mixed flora.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty