E.Z.N.A.® Food DNA Kit, Omega Bio-Tek
Supplier: Bierbaum-Proenen GmbH & Co.KG
D4616-01 D4616-00
75832-848EA 199.54 USD InStock
75832-848 75832-850
E.Z.N.A.® Food DNA Kit, Omega Bio-Tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Food DNA Kit allows rapid and reliable isolation of high-quality DNA from complex matrixes such as processed food, chocolate, cereals and meat.
  • Allows for efficient homogenization of samples
  • Eliminate PCR inhibiting compounds within the samples and elute highly concentrated DNA
  • No organic extractions
The E.Z.N.A.® Food DNA Kit allows rapid and reliable isolation of high-quality DNA from complex matrixes such as processed food, chocolate, cereals and meat. Specific protocols exist for target host DNA for GMO testing or bacterial DNA for pathogen/spoilation testing. Omega Bio-tek’s MB1 Buffer allows for efficient homogenization of samples without foaming commonly seen in lysis buffers containing detergents resulting in higher yields. The system combines the Omega Bio-tek’s HiBind® DNA Mini Columns with RBB Buffer to eliminate PCR inhibiting compounds within the samples and elute highly concentrated DNA. The purified DNA can be used in PCR-based testing for genetically modified organism (GMO) DNA. There are no organic extractions thus reducing plastic waste and hands-on time and multiple samples can be processed in parallel.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty