Carboxylic acid resin (MP), beads_macroporous 330-1225 µm, 15-50 mesh (>90% within) Loading: 3.4-3.6 mmol/g
Supplier: ESRI Magyarország Kft.
SPMP29-100 SPMP29-1KG
103061-878EA 98 USD InStock
103061-878 103061-880
Carboxylic acid resin (MP), beads_macroporous 330-1225 µm, 15-50 mesh (>90% within) Loading: 3.4-3.6 mmol/g
Carboxylic acid resin
Solid supported carboxylic acid utilized in solid phase synthesis for coupling amino acids at amino terminus.
Carboxylic Acid Resin, MP, is a macroporous resin functionalized with a carboxylic acid acid end group. It is a weak cation exchange resin, and has a different scavenging profile than MP-TsOH (SCX) due to its weaker acidity. The resin is designed to perform well in affinity chromatography based separation of peptides and proteins under aqueous elution conditions.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9177a879fd054fbd1317903d68502359302eceee&vid=9177a879fd054fbd1317903d68502359302eceee&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Colour Off-white
Form Granule
Loading 3.4-3.6 mmol/g
Particle size 330-1225 μm (15-50 mesh)
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty