Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diisopropylethylamine resin (MP), beads_macroporous 330-1225 µm, 15-50 mesh(>90% within) Loading: 2.4-2.8 mmol/g

Supplier: ESRI Magyarország Kft.

SPMP10-25 SPMP10-100 SPMP10-1KG
103062-016EA 93.85 USD InStock
103062-016 103062-018 103062-020
Diisopropylethylamine resin (MP), beads_macroporous 330-1225 µm, 15-50 mesh(>90% within) Loading: 2.4-2.8 mmol/g
Diisopropylethylamine resin
Solid supported tertiary amine useful in all acid scavenging protocols.

Diisopropylethylamine Resin, MP, is a macroporous resin functionalized with a diisopropyl end group and is functionally equivalent to Huenig’s Base. It is very effective in reactions that produce acidic byproducts. MP-DIEA is extremely efficient in sequestering HCl formed during amide, sulfonamide and carbamate synthesis. When it is combined with MP-Trisamine (scavenges excess acid, sulfonyl and carbamoyl chlorides) the result is a one-pot synthesis, which requires only filtration to isolate the desired product. The steric bulk associated with the two isopropyl groups severely inhibits the nucleophilicity of the amine towards electrophilic partners, therefore removing the possibility of reaction byproducts arising from the resin itself.


https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f773d027d3630a4bdd92bfc35097aa2e0138f71&vid=7f773d027d3630a4bdd92bfc35097aa2e0138f71&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Colour Off-white
Form Granule
Loading 1.2-1.4 mmol/g
Particle size 330-1225 μm (15-50 mesh)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty