BBL™ Selenite Broth, BD
Supplier: Seca Med.Waagen u.Meßsysteme
221020 221021
90001-190PK 35.7 USD InStock
90001-190 90001-192 90004-976
BBL™ Selenite Broth, BD
Media for Microbiology Prepared Media for Microbiology
Selenite Cystine Broth is used as a selective enrichment medium for the isolation of Salmonella from feces, foods, water, and other materials of sanitary importance.
  • Peptone provides amino acids and nitrogen
  • Lactose is a source of energy
  • Sodium phosphate buffers medium to maintain pH level
  • Sodium selenite inhibits gram-positive bacteria and most gram-negative enterics
  • L-cystine is incorporated to improve the recovery of Salmonella
Selenite Cystine Broth and similar enrichment media (Selenite-F Broth) are useful for detecting Salmonella in the nonacute stages of illness when the organisms occur in the feces in low numbers and for epidemiological studies to enhance the detection of low numbers of organisms from asymptomatic or convalescent patients.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty