Adhesive Vinyl GHS OSHA Labels 'Ethylene Glycol', HCL Labels
Supplier: HCL Label
GH10059VN0023 GH10059VN0035 GH10059VN0047 GH20059VN0023 GH20059VN0035 GH20059VN0047 GH30059VN0023 GH30059VN0035 GH30059VN0047 GH40059VN0023 GH40059VN0035 GH40059VN0047
103019-646PK 74.03 USD InStock
103019-646 103019-648 103019-650 103019-652 103019-654 103019-656 103027-558 103027-560 103027-562 103027-564 103027-566 103027-568 103035-352 103035-354 103035-356 103035-358 103035-360 103035-362 103043-468 103043-470 103043-472 103043-474 103043-556 103043-558
Adhesive Vinyl GHS OSHA Labels 'Ethylene Glycol', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certificaciones: RoHS compliant
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty