Adhesive Vinyl GHS OSHA Labels 'Lapping Solution', HCL Labels
Supplier: HCL Label
GH10183VN0023 GH10183VN0035 GH10183VN0047 GH20183VN0023 GH20183VN0035 GH20183VN0047 GH30183VN0023 GH30183VN0035 GH30183VN0047 GH40183VN0023 GH40183VN0035 GH40183VN0047
103020-134PK 74.03 USD InStock
103020-134 103020-136 103020-138 103020-140 103020-142 103020-144 103027-934 103027-936 103027-938 103027-940 103027-942 103027-944 103035-248 103035-250 103035-252 103035-254 103035-808 103035-810 103043-806 103043-868 103043-870 103043-872 103043-874 103043-876
Adhesive Vinyl GHS OSHA Labels 'Lapping Solution', HCL Labels
Labels
These laminated ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certificaciones: RoHS compliant.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty